2-Amino-4-fluorobenzenesulphonamide

Supplier: APOLLO SCIENTIFIC
Danger

PC501625-500MG PC501625-1G PC501625-250MG
77416-114EA 407.52 USD
77416-114 77416-110 77416-112
2-Amino-4-fluorobenzenesulphonamide
2-Amino-4-fluorobenzenesulphonamide
Formula: C₆H₇FN₂O₂S
MDL Number: MFCD11125174
CAS Number: 2822-72-2
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR