(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)boronic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,2-Difluoro-1,3-benzodioxole-5-boronic acid, (2,2-Difluorobenzo[d][1,3]dioxol-5-yl)boronic acid

PC501601-1G PC501601-25G PC501601-5G
77416-098EA 87.62 USD
77416-098 77416-100 77416-102
(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)boronic acid 98%
(2,2-Difluorobenzo[d][1,3]dioxol-5-yl)boronic acid
Formula: C₇H₅BF₂O₄
MW: 201.92 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08706236
CAS Number: 190903-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR