4-Bromo-5-fluoro-2-iodobenzyl bromide 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-Bromo-4-(bromomethyl)-2-fluoro-5-iodobenzene

PC501571-250MG PC501571-5G PC501571-1G
77416-094EA 166.06 USD
77416-094 77416-096 77416-092
4-Bromo-5-fluoro-2-iodobenzyl bromide 95%
4-Bromo-5-fluoro-2-iodobenzyl bromide
Formula: C₇H₄Br₂FI
MW: 393.82 g/mol
MDL Number: MFCD18910205
CAS Number: 1379330-38-7
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR