2-Bromo-5-fluoro-3-(trifluoromethyl)phenol 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC501305-1G PC501305-250MG PC501305-5G
77416-050EA 484.96 USD
77416-050 77416-052 77416-054
2-Bromo-5-fluoro-3-(trifluoromethyl)phenol 95%
2-Bromo-5-fluoro-3-(trifluoromethyl)phenol
Formula: C₇H₃BrF₄O
MDL Number: MFCD28348577
CAS Number: 1807145-12-5
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR