4-Bromo-3-fluoro-2-methoxybenzonitrile 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC500667-1G PC500667-250MG
77416-020EA 399.38 USD
77416-020 77416-022
4-Bromo-3-fluoro-2-methoxybenzonitrile 98%
4-Bromo-3-fluoro-2-methoxybenzonitrile
Formula: C₈H₅BrFNO
MDL Number: MFCD23707400
CAS Number: 1427452-84-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR