2-Fluoro-4-methoxy-5-nitrobenzyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

PC500448-1G PC500448-5G
77416-010EA 578.68 USD
77416-010 77416-012
2-Fluoro-4-methoxy-5-nitrobenzyl bromide
2-Fluoro-4-methoxy-5-nitrobenzyl bromide
Formula: C₈H₇BrFNO₃
MW: 264.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27995570
CAS Number: 618914-29-7
UN: 3261

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR