4'-Bromo-2,2,2,2',3'-pentafluoroacetophenone 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC500212-1G PC500212-250MG
77416-002EA 616.38 USD
77416-002 77416-004
4'-Bromo-2,2,2,2',3'-pentafluoroacetophenone 95%
4'-Bromo-2,2,2,2',3'-pentafluoroacetophenone
Formula: C₈H₂BrF₅O
MW: 289 g/mol
MDL Number: MFCD27664852
CAS Number: 1823361-42-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR