2-Amino-4-fluoro-3-(trifluoromethyl)benzonitrile 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 6-Cyano-3-fluoro-2-(trifluoromethyl)aniline, 4-Fluoro-3-(trifluoromethyl)anthranilonitrile, 2-Amino-3-cyano-6-fluorobenzotrifluoride

PC49369-1G PC49369-250MG
77415-922EA 392.24 USD
77415-922 77415-924
2-Amino-4-fluoro-3-(trifluoromethyl)benzonitrile 95%
2-Amino-4-fluoro-3-(trifluoromethyl)benzonitrile
Formula: C₈H₄F₄N₂
MW: 204.12 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24850042
CAS Number: 1440535-22-7
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR