5-Bromo-2-chloro-4-fluorobenzonitrile 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC47466-1G PC47466-250MG PC47466-5G
77415-690EA 350.48 USD
77415-690 77415-692 77415-694
5-Bromo-2-chloro-4-fluorobenzonitrile 95%
5-Bromo-2-chloro-4-fluorobenzonitrile
Formula: C₇H₂BrClFN
CAS Number: 1892285-40-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR