2-Fluorothiophenol 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Fluorobenzenethiol, 1-Fluoro-2-mercaptobenzene, 2-Fluorobenzene-1-thiol, o-Fluorothiophenol

PC4298-100G PC4298-25G PC4298-5G
77415-238EA 206.82 USD
77415-238 77415-240 77415-242
2-Fluorothiophenol 98%
2-Fluorothiophenol
Formula: C₆H₅FS
MW: 128.17 g/mol
Boiling Pt: 61…62 °C/35mm
Density: 1.203
Flash Pt: 56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041419
CAS Number: 2557-78-0
UN: 3336
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR