2,3-Difluoro-1,4-benzenediol 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,3-Difluorohydroquinone

PC408062-1G PC408062-250MG
77415-094EA 139.58 USD
77415-094 77415-096
2,3-Difluoro-1,4-benzenediol 97%
2,3-Difluoro-1,4-benzenediol
Formula: C₆H₄F₂O₂
MW: 146.092 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04038791
CAS Number: 124728-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR