4-Fluorobenzamidine hydrochloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC401011-10G PC401011-1G PC401011-5G
77415-056EA 92.72 USD
77415-056 77415-058 77415-060
4-Fluorobenzamidine hydrochloride 97%
4-Fluorobenzamidine hydrochloride
Formula: C₇H₈ClFN₂
Melting Pt: 221 °C
MDL Number: MFCD04114421
CAS Number: 456-14-4
UN: 2811

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR