2,5-Difluoro-4-(trifluoromethyl)benzyl bromide 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-(Bromomethyl)-2,5-difluoro-4-(trifluoromethyl)benzene

PC34215-1G PC34215-250MG
77414-726EA 275.08 USD
77414-726 77414-728
2,5-Difluoro-4-(trifluoromethyl)benzyl bromide 95%
2,5-Difluoro-4-(trifluoromethyl)benzyl bromide
Formula: C₈H₄BrF₅
MW: 275.01 g/mol
MDL Number: MFCD34181543
CAS Number: 2385579-27-9
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR