2,5-Difluoro-4-(trifluoromethyl)benzyl alcohol 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: [2,5-Difluoro-4-(trifluoromethyl)phenyl]methanol

PC33177-1G PC33177-250MG PC33177-5G
77414-688EA 206.82 USD
77414-688 77414-690 77414-692
2,5-Difluoro-4-(trifluoromethyl)benzyl alcohol 95%
2,5-Difluoro-4-(trifluoromethyl)benzyl alcohol
Formula: C₈H₅F₅O
MDL Number: MFCD28063904
CAS Number: 1803598-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR