4-Bromo-2-fluoro-5-(trifluoromethyl)benzyl bromide 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC305030-1G PC305030-250MG
77414-426EA 619.44 USD
77414-426 77414-428
4-Bromo-2-fluoro-5-(trifluoromethyl)benzyl bromide 95%
4-Bromo-2-fluoro-5-(trifluoromethyl)benzyl bromide
Formula: C₈H₄Br₂F₄
MW: 335.92 g/mol
MDL Number: MFCD30748835
CAS Number: 2092187-20-5
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR