4-Bromo-2-fluoro-5-(trifluoromethyl)benzonitrile 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC305029-1G PC305029-250MG PC305029-5G
77414-420EA 496.16 USD
77414-420 77414-422 77414-424
4-Bromo-2-fluoro-5-(trifluoromethyl)benzonitrile 95%
4-Bromo-2-fluoro-5-(trifluoromethyl)benzonitrile
Formula: C₈H₂BrF₄N
MW: 268.01 g/mol
MDL Number: MFCD32265190
CAS Number: 2091422-93-2
UN: 2811
ADR: 6.1 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR