Ethyl 7-(trifluoromethoxy)-1-benzofuran-2-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

PC301061-1G PC301061-250MG
77414-086EA 426.88 USD
77414-086 77414-088
Ethyl 7-(trifluoromethoxy)-1-benzofuran-2-carboxylate
Ethyl 7-(trifluoromethoxy)-1-benzofuran-2-carboxylate
Formula: C₁₂H₉F₃O₄
MDL Number: MFCD34596998
CAS Number: 896732-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR