2-Chloro-5-(trifluoromethyl)phenol 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC1930W-100G PC1930W-25G PC1930W-5G
77413-420EA 131.42 USD
77413-420 77413-422 77413-424
2-Chloro-5-(trifluoromethyl)phenol 98%
2-Chloro-5-(trifluoromethyl)phenol
Formula: C₇H₄ClF₃O
Boiling Pt: 87…88 °C
Density: 1.459
Flash Pt: 46 °C
MDL Number: MFCD00019995
CAS Number: 40889-91-6
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR