1-bromo-4-(bromomethyl)-2-fluoro-3-methoxybenzene

Supplier: APOLLO SCIENTIFIC
Danger

PC104146-1G PC104146-250MG PC104146-5G
77412-870EA 756.98 USD
77412-870 77412-872 77412-874
1-bromo-4-(bromomethyl)-2-fluoro-3-methoxybenzene
1-bromo-4-(bromomethyl)-2-fluoro-3-methoxybenzene
Formula: C₈H₇Br₂FO
MW: 297.95 g/mol
CAS Number: 2090318-84-4
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR