4-bromo-5-fluoro-2-methoxybenzyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

PC104145-1G PC104145-5G
77412-866EA 578.68 USD
77412-866 77412-868
4-bromo-5-fluoro-2-methoxybenzyl bromide
4-bromo-5-fluoro-2-methoxybenzyl bromide
Formula: C₈H₇Br₂FO
MW: 297.95 g/mol
Storage Temperature: Refrigerator
CAS Number: 1823875-73-5
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR