5-bromo-2-(bromomethyl)-1-fluoro-3-methylbenzene

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-Bromo-4-(bromomethyl)-3-fluoro-5-methyl-benzene

PC104123-1G PC104123-5G
77412-856EA 496.16 USD
77412-856 77412-858
5-bromo-2-(bromomethyl)-1-fluoro-3-methylbenzene
5-bromo-2-(bromomethyl)-1-fluoro-3-methylbenzene
Formula: C₈H₇Br₂F
MW: 281.95 g/mol
MDL Number: MFCD34180132
CAS Number: 2091144-14-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR