5-Bromo-2-fluoro-4-iodobenzaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

PC104102-250MG PC104102-5G PC104102-1G
77412-834EA 273.04 USD
77412-834 77412-836 77412-832
5-Bromo-2-fluoro-4-iodobenzaldehyde
5-Bromo-2-fluoro-4-iodobenzaldehyde
Formula: C₇H₃BrFIO
MDL Number: MFCD30639897
CAS Number: 2092699-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR