5-Bromo-2-fluoro-4-iodobenzyl bromide

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-Bromo-5-(bromomethyl)-4-fluoro-2-iodobenzene

PC104091-1G PC104091-5G
77412-828EA 578.68 USD
77412-828 77412-830
5-Bromo-2-fluoro-4-iodobenzyl bromide
5-Bromo-2-fluoro-4-iodobenzyl bromide
Formula: C₇H₄Br₂FI
MW: 393.82 g/mol
CAS Number: 2091400-72-3
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR