5-Bromo-2-fluoro-4-iodobenzyl alcohol

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (5-Bromo-2-fluoro-4-iodophenyl)methanol

PC104066-1G PC104066-5G
77412-824EA 578.68 USD
77412-824 77412-826
5-Bromo-2-fluoro-4-iodobenzyl alcohol
5-Bromo-2-fluoro-4-iodobenzyl alcohol
Formula: C₇H₅BrFIO
MDL Number: MFCD33401574
CAS Number: 2092124-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR