3-Bromo-2-hydroxy-5-iodobenzaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

OR97136-1G OR97136-25G OR97136-5G
77412-282EA 544.04 USD
77412-282 77412-284 77412-286
3-Bromo-2-hydroxy-5-iodobenzaldehyde
3-Bromo-2-hydroxy-5-iodobenzaldehyde
Formula: C₇H₄BrIO₂
MW: 326.91 g/mol
MDL Number: MFCD29921859
CAS Number: 1582226-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR