5-Chloro[1,2,4]triazolo[1,5-c]pyrimidine 96%

Supplier: APOLLO SCIENTIFIC
Warning

OR960645-100MG OR960645-1G OR960645-500MG
77412-222EA 114.1 USD
77412-222 77412-224 77412-226
5-Chloro[1,2,4]triazolo[1,5-c]pyrimidine 96%
5-Chloro[1,2,4]triazolo[1,5-c]pyrimidine
Formula: C₅H₃ClN₄
Storage Temperature: Freezer
MDL Number: MFCD09839072
CAS Number: 76044-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR