2-Chloro-9-methyl-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carbaldehyde 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR946315-1G OR946315-250MG OR946315-5G
77412-124EA 170.14 USD
77412-124 77412-126 77412-128
2-Chloro-9-methyl-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carbaldehyde 95%
2-Chloro-9-methyl-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carbaldehyde
Formula: C₁₀H₇ClN₂O₂
MW: 222.62 g/mol
MDL Number: MFCD03284432
CAS Number: 17326-27-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR