4-Bromo-2-isopropoxypyridine 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR943968-250MG OR943968-5G OR943968-1G
77412-114EA 124.3 USD
77412-114 77412-116 77412-112
4-Bromo-2-isopropoxypyridine 98%
4-Bromo-2-isopropoxypyridine
Formula: C₈H₁₀BrNO
Density: 1.383
MDL Number: MFCD11869614
CAS Number: 1142194-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR