3-Methyl-4-nitrobenzyl bromide 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR927531-100G OR927531-25G OR927531-5G
77411-904EA 1008.62 USD
77411-904 77411-906 77411-908
3-Methyl-4-nitrobenzyl bromide 98%
3-Methyl-4-nitrobenzyl bromide
Formula: C₈H₈BrNO₂
Melting Pt: 46…48 °C
MDL Number: MFCD04039920
CAS Number: 141281-38-1
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR