Dibutyl sulphate 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Di-n-butylsulphate

OR924973-100G OR924973-25G OR924973-5G
77411-890EA 261.84 USD
77411-890 77411-892 77411-894
Dibutyl sulphate 95%
Dibutyl sulphate
Formula: C₈H₁₈O₄S
MW: 210.29 g/mol
Boiling Pt: 115…116 °C
Density: 1.06
MDL Number: MFCD00059420
CAS Number: 625-22-9
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR