(R)-Fmoc-2-amino-3-ethyl-pentanoic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR923122-1G OR923122-250MG
77411-826EA 1128.84 USD
77411-826 77411-828
(R)-Fmoc-2-amino-3-ethyl-pentanoic acid
(R)-Fmoc-2-amino-3-ethyl-pentanoic acid
Formula: C₂₂H₂₅NO₄
MDL Number: MFCD12408089
CAS Number: 1310680-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR