Methyl 2-amino-6-bromobenzoate 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR916421-1G OR916421-250MG OR916421-5G
77411-696EA 81.5 USD
77411-696 77411-698 77411-700
Methyl 2-amino-6-bromobenzoate 97%
Methyl 2-amino-6-bromobenzoate
Formula: C₈H₈BrNO₂
MW: 230.06 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11977360
CAS Number: 135484-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR