4-Chloro-4'-methoxybenzophenone 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR912631-1G OR912631-5G
77411-666EA 83.54 USD
77411-666 77411-668
4-Chloro-4'-methoxybenzophenone 98%
4-Chloro-4'-methoxybenzophenone
Formula: C₁₄H₁₁ClO₂
MDL Number: MFCD01105071
CAS Number: 10547-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR