(R)-1-Chloropropan-2-ol 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: (R)-1-Chloro-2-propanol

OR911848-1G OR911848-250MG OR911848-5G
77411-626EA 103.92 USD
77411-626 77411-628 77411-630
(R)-1-Chloropropan-2-ol 98%
(R)-1-Chloropropan-2-ol
Formula: C₃H₇ClO
MW: 94.54 g/mol
MDL Number: MFCD01862132
CAS Number: 19141-39-0
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR