2-Amino-6-chloro-4-nitrophenol 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR903127-100G OR903127-25G OR903127-5G
77411-530EA 364.74 USD
77411-530 77411-532 77411-534
2-Amino-6-chloro-4-nitrophenol 95%
2-Amino-6-chloro-4-nitrophenol
Formula: C₆H₅ClN₂O₃
Melting Pt: 160…164 °C
Storage Temperature: Freezer
MDL Number: MFCD00035767
CAS Number: 6358-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR