2,4,6-Trimethyl-m-phenylenediamine
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
1,3-Diamino-2,4,6-trimethylbenzene, 2,4-Diamino-1,3,5-trimethylbenzene, 2,4,6-Trimethyl-1,3-phenylenediamine, 2,4-Diaminomesitylene, 2,4,6-Trimethylbenzene-1,3-diamine
OR903076-100G
OR903076-25G
77411-526EA
170.16
USD
77411-526
77411-528
2,4,6-Trimethyl-m-phenylenediamine
2,4,6-Trimethyl-m-phenylenediamine
Formula:
C₉H₁₄N₂ Melting Pt: 89…91 °C |
MDL Number:
MFCD00010149 CAS Number: 3102-70-3 UN: 3077 ADR: 9+EHS,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...