2,4-Dichloro-5-nitrophenol 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1,3-Dichloro-4-hydroxy-6-nitrobenzene, 2,4-Dichloro-5-nitrobenzenol

OR8241-1G OR8241-5G
77411-376EA 69.28 USD
77411-376 77411-378
2,4-Dichloro-5-nitrophenol 98%
2,4-Dichloro-5-nitrophenol
Formula: C₆H₃Cl₂NO₃
MDL Number: MFCD03093072
CAS Number: 39489-77-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR