Ethyl oxo(2-oxocyclopent-1-yl)acetate 90%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Ethyl 2-oxo-2-(2-oxocyclopentyl)acetate, Ethyl (2-oxocyclopent-1-yl)glyoxylate, Ethyl 2-oxo-2-(2-oxocyclopent-1-yl)ethanoate

OR72361-1G OR72361-5G
77411-188EA 47.84 USD
77411-188 77411-190
Ethyl oxo(2-oxocyclopent-1-yl)acetate 90%
Ethyl oxo(2-oxocyclopent-1-yl)acetate
Formula: C₉H₁₂O₄
Boiling Pt: 129…130 °C
Melting Pt: 26…31 °C
Density: 1.37
MDL Number: MFCD00019311
CAS Number: 39163-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR