Dicyclohexylphenylphosphine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Dicyclohexyl(phenyl)phosphine

OR72334-1G OR72334-5G
77411-166EA 58.08 USD
77411-166 77411-168
Dicyclohexylphenylphosphine
Dicyclohexylphenylphosphine
Formula: C₁₈H₂₇P
MW: 274.38 g/mol
Melting Pt: 61…61 °C
MDL Number: MFCD00003854
CAS Number: 6476-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR