4-(Chlorosulphonyl)-3-nitrobenzoic acid

Supplier: APOLLO SCIENTIFIC
Danger

OR72330-1G OR72330-5G
77411-162EA 441.14 USD
77411-162 77411-164
4-(Chlorosulphonyl)-3-nitrobenzoic acid
4-(Chlorosulphonyl)-3-nitrobenzoic acid
Formula: C₇H₄ClNO₆S
MW: 265.63 g/mol
Melting Pt: 198…199 °C
MDL Number: MFCD06254510
CAS Number: 32571-65-6
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR