2-((Tetrahydro-2H-pyran-2-yl)oxy)ethanol 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR72321-1G OR72321-25G OR72321-5G
77411-156EA 89.66 USD
77411-156 77411-158 77411-160
2-((Tetrahydro-2H-pyran-2-yl)oxy)ethanol 95%
2-((Tetrahydro-2H-pyran-2-yl)oxy)ethanol
Formula: C₇H₁₄O₃
Boiling Pt: 95 °C
Density: 1.077
Flash Pt: 108 °C
MDL Number: MFCD00185588
CAS Number: 2162-31-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR