Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II)

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: [4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene]palladium(II) Dichloride, [4,5-Bis(diphenylphosphino)-9,9-dimethylxanthene]dichloropalladium(II)

OR72312-5G OR72312-1G
77411-154EA 172.18 USD
77411-154 77411-152
Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II)
Dichloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene]palladium(II)
Formula: C₃₉H₃₂Cl₂OP₂Pd
MW: 755.95 g/mol
Melting Pt: 287 °C
MDL Number: MFCD14155707
CAS Number: 205319-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR