Bis(triphenylphosphine)palladium(II) diacetate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Bis(triphenylphosphine)palladium(II) acetate, Bis(acetato)bis(triphenylphosphine)palladium(II), Diacetatobis(triphenylphosphine)palladium(II)

OR72310-10G OR72310-1G OR72310-5G
77411-146EA 303.6 USD
77411-146 77411-148 77411-150
Bis(triphenylphosphine)palladium(II) diacetate
Bis(triphenylphosphine)palladium(II) diacetate
Formula: C₄₀H₃₆O₄P₂Pd
Melting Pt: 136 °C
MDL Number: MFCD00010013
CAS Number: 14588-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR