3-(Phenylthio)benzyl acrylate

Supplier: APOLLO SCIENTIFIC
Warning

OR72286-1G OR72286-250MG OR72286-5G
77411-134EA 578.68 USD
77411-134 77411-136 77411-138
3-(Phenylthio)benzyl acrylate
3-(Phenylthio)benzyl acrylate
Formula: C₁₆H₁₄O₂S
CAS Number: 2170636-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR