Methyl 3-(bromomethyl)-4-methoxybenzoate

Supplier: APOLLO SCIENTIFIC
Danger

OR72282-1G OR72282-250MG OR72282-5G
77411-128EA 253.68 USD
77411-128 77411-130 77411-132
Methyl 3-(bromomethyl)-4-methoxybenzoate
Methyl 3-(bromomethyl)-4-methoxybenzoate
Formula: C₁₀H₁₁BrO₃
MW: 259.1 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14706047
CAS Number: 142031-88-7
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR