3-(4-Chlorophenyl)pentanedioic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR72262-100G OR72262-25G OR72262-500G
77411-116EA 110.04 USD
77411-116 77411-118 77411-120
3-(4-Chlorophenyl)pentanedioic acid 97%
3-(4-Chlorophenyl)pentanedioic acid
Formula: C₁₁H₁₁ClO₄
Melting Pt: 164…166 °C
MDL Number: MFCD00190249
CAS Number: 35271-74-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR