(1-Bromo-3-(bromomethyl)-2-chloro-5-iodobenzene)

Supplier: APOLLO SCIENTIFIC
Danger

OR72250-1G OR72250-5G
77411-110EA 29.54 USD
77411-110 77411-112
(1-Bromo-3-(bromomethyl)-2-chloro-5-iodobenzene)
(1-Bromo-3-(bromomethyl)-2-chloro-5-iodobenzene)
Formula: C₇H₄Br₂ClI
MDL Number: MFCD34603431
CAS Number: 2090530-81-5
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR