Nicotinic acid hydrazide tech

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Nicotinohydrazide, (3-Pyridylcarbonyl)hydrazine, Pyridine-3-carbohydrazide

OR72208-100G OR72208-25G
77411-104EA 54.46 USD
77411-104 77411-106
Nicotinic acid hydrazide tech
Nicotinic acid hydrazide
Formula: C₆H₇N₃O
Melting Pt: 159…161 °C
MDL Number: MFCD00006383
CAS Number: 553-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR