Methyl 3-amino-6-bromopicolinate

Supplier: APOLLO SCIENTIFIC
Warning

OR72204-1G OR72204-5G
77411-100EA 230.26 USD
77411-100 77411-102
Methyl 3-amino-6-bromopicolinate
Methyl 3-amino-6-bromopicolinate
Formula: C₇H₇BrN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD11656228
CAS Number: 866775-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR