Lithium triisopropoxy(thiazol-2-yl)borate

Supplier: APOLLO SCIENTIFIC
Warning

OR72200-100MG OR72200-1G OR72200-250MG
77411-094EA 63.16 USD
77411-094 77411-096 77411-098
Lithium triisopropoxy(thiazol-2-yl)borate
Lithium triisopropoxy(thiazol-2-yl)borate
Formula: C₁₂H₂₃BLiNO₃S
MW: 279.13 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16872058
CAS Number: 1393823-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR